2,4,6-Trichlorpyridin ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
H56787.06
H56787.14
H56787.06EA
111
EUR
H56787.06
H56787.14
2,4,6-Trichlorpyridin ≥97%
2,4,6-Trichlorpyridin
Purity: 97%
CAS No.: 16063-69-7
Molecular Formula: C5H2Cl3N
Formula Weight: 182.44
Flash Point:
Formel:
C₅H₂Cl₃N Molecular Weight: 182,44 g/mol Schmelzpunkt: 33…37 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00971928 CAS-Nummer: 16063-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...