2-Brom-α,α,α,4-tetrafluortoluol
Lieferant: COMBI-BLOCKS
Synonyme:
1-Brom-3-fluor-6-(trifluormethyl)benzol, 2-Brom-4-fluorbenzotrifluorid
OS-7725-1G
OS-7725-5G
OS-7725-25G
COBBOS-7725-1GEA
26.2
EUR
COBBOS-7725-1G
COBBOS-7725-5G
COBBOS-7725-25G
2-Brom-α,α,α,4-tetrafluortoluol
2-Brom-α,α,α,4-tetrafluortoluol
Formel:
C₇H₃BrF₄ Molecular Weight: 243 g/mol |
MDL:
MFCD03094041 CAS-Nummer: 351003-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...