2,3,4-Trifluorphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 2,3,4-Trifluorbenzolboronsäure, (2,3,4-Trifluorphenyl)metaborsäure, Dihydroxy(2,3,4-trifluorphenyl)boran

016268-1G 016268-5G
FLUO016268-1GEA 31.1 EUR
FLUO016268-1G FLUO016268-5G
2,3,4-Trifluorphenylboronsäure
2,3,4-Trifluorphenylboronsäure
Formel: C₆H₄BF₃O₂
Molecular Weight: 175,9 g/mol
MDL: MFCD01863168
CAS-Nummer: 226396-32-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR