5-Brom-2,3-dichlorpyridin
Lieferant: FLUOROCHEM
012745-1G
012745-5G
FLUO012745-1GEA
31.1
EUR
FLUO012745-1G
FLUO012745-5G
5-Brom-2,3-dichlorpyridin
5-Brom-2,3-dichlorpyridin
Formel:
C₅H₂BrCl₂N Molecular Weight: 226.89 g/mol |
MDL:
MFCD04112492 CAS-Nummer: 97966-00-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...