2-Fluor-5-(trifluormethyl)benzylalkohol

Lieferant: FLUOROCHEM

Synonyme: (2-Fluoro-5-(trifluoromethyl)phenyl)methanol

006411-1G 006411-5G 006411-10G
FLUO006411-1GEA 31.1 EUR
FLUO006411-1G FLUO006411-5G FLUO006411-10G
2-Fluor-5-(trifluormethyl)benzylalkohol
2-Fluor-5-(trifluormethyl)benzylalkohol
Formel: C₈H₆F₄O
Molecular Weight: 194.13 g/mol
MDL: MFCD00061167
CAS-Nummer: 207974-09-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR