3,6-Dibrom-2-picolin

Lieferant: FLUOROCHEM

Synonyme: 3,6-Dibrom-2-methylpyridin

012710-1G 012710-5G
FLUO012710-1GEA 31.1 EUR
FLUO012710-1G FLUO012710-5G
3,6-Dibrom-2-picolin
3,6-Dibrom-2-picolin
Formel: C₆H₅Br₂N
Molecular Weight: 250,92 g/mol
MDL: MFCD06254589
CAS-Nummer: 39919-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR