4-Iodbenzonitril

Lieferant: FLUOROCHEM

Synonyme: p-Iodobenzonitrile

147902-25G 147902-5G
FLUO147902-25GEA 78.9 EUR
FLUO147902-25G FLUO147902-5G
4-Iodbenzonitril
4-Iodbenzonitril
Formel: C₇H₄IN
Molecular Weight: 229,02 g/mol
Siedepunkt: 285 °C (1013 hPa)
Schmelzpunkt: 125…128 °C
MDL: MFCD00051310
CAS-Nummer: 3058-39-7
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR