(2-Nitro-4-(trifluormethyl)phenyl)essigsäure

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 2-[2-Nitro-4-(trifluoromethyl)phenyl]acetic acid

CA-4360-5G CA-4360-25G CA-4360-1G
COBBCA-4360-5GEA 125 EUR
COBBCA-4360-5G COBBCA-4360-25G COBBCA-4360-1G
(2-Nitro-4-(trifluormethyl)phenyl)essigsäure
(2-Nitro-4-(trifluormethyl)phenyl)essigsäure
Formel: C₉H₆F₃NO₄
Molecular Weight: 249.15 g/mol
MDL: MFCD00091534
CAS-Nummer: 1735-91-7
EINECS: 217-081-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR