3-(Methylthio)phenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 3-Thioanisoleboronic acid, 3-(Methylthio)benzeneboronic acid

016264-1G 016264-5G
FLUO016264-1GEA 31.1 EUR
FLUO016264-1G FLUO016264-5G
3-(Methylthio)phenylboronsäure
3-(Methylthio)phenylboronsäure
Formel: C₇H₉BO₂S
MDL: MFCD01319105
CAS-Nummer: 128312-11-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR