Boc-DL-Pro-OH ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid, N-Boc-DL-proline

H32347.03 H32347.06
H32347.03EA 56.1 EUR
H32347.03 H32347.06
Boc-DL-Pro-OH ≥98%
Boc-DL-Pro-OH

Purity: 98% CAS No.: 59433-50-0 Molecular Formula: C10H17NO4 Formula Weight: 215.25 Melting Point: 101-104° MDL No.: MFCD00520915

Formel: C₁₀H₁₇NO₄
Molecular Weight: 215.25 g/mol
Schmelzpunkt: 101…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00520915
CAS-Nummer: 59433-50-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR