Benzofurazan
Lieferant: Thermo Scientific
Synonyme: 2,1,3-Benzoxadiazole, Benzo[c][1,2,5]oxadiazole
KM06477.250 KM06477DA
MAYBKM06477.250EA 0 EUR
MAYBKM06477.250 MAYBKM06477DA
Benzofurazan
Benzofurazan
Formel: C₆H₄N₂O
Molecular Weight: 120,11 g/mol
MDL: MFCD00068064
CAS-Nummer: 273-09-6