5-Brompicolinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromopyridine-2-carboxylic acid, 5-Bromo-2-pyridinecarboxylic acid

CA-4116-25G CA-4116-5G CA-4116-100G
COBBCA-4116-25GEA 171 EUR
COBBCA-4116-25G COBBCA-4116-5G COBBCA-4116-100G
5-Brompicolinsäure
5-Brompicolinsäure
Formel: C₆H₄BrNO₂
Molecular Weight: 202.01 g/mol
MDL: MFCD00234149
CAS-Nummer: 30766-11-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR