(S)-(-)-1,1'-Bi-2-naphthol

Lieferant: COMBI-BLOCKS

Synonyme: (S)-BINOL, (S)-(-)-1,1'-Binaphthyl-2,2'-Diol, (S)-[1,1'-Binaphthalene]-2,2'-diol

CS-9131-5G CS-9131-25G CS-9131-100G
COBBCS-9131-5GEA 25.5 EUR
COBBCS-9131-5G COBBCS-9131-25G COBBCS-9131-100G
(S)-(-)-1,1'-Bi-2-naphthol
(S)-(-)-1,1'-Bi-2-naphthol
Formel: C₂₀H₁₄O₂
Molecular Weight: 286,33 g/mol
MDL: MFCD00004068
CAS-Nummer: 18531-99-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR