Methyl-2-amino-4-ethyl-5-methyl-3-thiophencarboxylat
Lieferant: FLUOROCHEM
014025-1G
014025-5G
FLUO014025-1GEA
171
EUR
FLUO014025-1G
FLUO014025-5G
Methyl-2-amino-4-ethyl-5-methyl-3-thiophencarboxylat
Methyl-2-amino-4-ethyl-5-methyl-3-thiophencarboxylat
Formel:
C₉H₁₃NO₂S |
MDL:
MFCD00781744 CAS-Nummer: 4815-25-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...