4-(Trifluormethoxy)toluol

Lieferant: COMBI-BLOCKS

OS-7659-100G OS-7659-5G OS-7659-1G OS-7659-25G
COBBOS-7659-100GEA 0 EUR
COBBOS-7659-100G COBBOS-7659-5G COBBOS-7659-1G COBBOS-7659-25G
4-(Trifluormethoxy)toluol
4-(Trifluormethoxy)toluol
Formel: C₈H₇F₃O
MDL: MFCD00042413
CAS-Nummer: 706-27-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR