(4-Nitrophenyl)-β-D-galactopyranosid ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
pNPG, 4-Nitrophenyl β-D-Galactopyranoside
H56905.06
H56905.09
H56905.06EA
680
EUR
H56905.06
H56905.09
(4-Nitrophenyl)-β-D-galactopyranosid ≥98%
(4-Nitrophenyl)-β-D-galactopyranosid
Purity: 98+%
CAS No.: 3150-24-1
Molecular Formula: C12H15NO8
Formula Weight: 301.25
Storage Temperature: -30°C to -10°C
Formel:
C₁₂H₁₅NO₈ Molecular Weight: 301.25 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00063256 CAS-Nummer: 3150-24-1 EINECS: 221-584-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...