1,1,2,3,3,3-Hexafluorpropyltrichlormethylether
Lieferant: FLUOROCHEM
010321-1G
010321-5G
FLUO010321-1GEA
82.1
EUR
FLUO010321-1G
FLUO010321-5G
1,1,2,3,3,3-Hexafluorpropyltrichlormethylether
1,1,2,3,3,3-Hexafluorpropyltrichlormethylether
Formel:
C₄HCl₃F₆O |
MDL:
MFCD03094226 CAS-Nummer: 56860-83-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...