1,1,2,3,3,3-Hexafluorpropyldifluormethylether
Lieferant: FLUOROCHEM
010320-1G
010320-5G
FLUO010320-1GEA
57.9
EUR
FLUO010320-1G
FLUO010320-5G
1,1,2,3,3,3-Hexafluorpropyldifluormethylether
1,1,2,3,3,3-Hexafluorpropyldifluormethylether
Formel:
C₄H₂F₈O |
MDL:
MFCD03094220 CAS-Nummer: 56860-85-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...