2,2,3,3,3-Pentafluorpropyldifluormethylether
Lieferant: FLUOROCHEM
Synonyme:
Difluoromethyl 2,2,3,3,3-pentafluoropropyl ether
010630-1G
010630-5G
FLUO010630-1GEA
0
EUR
FLUO010630-1G
FLUO010630-5G
2,2,3,3,3-Pentafluorpropyldifluormethylether
2,2,3,3,3-Pentafluorpropyldifluormethylether
Formel:
C₄H₃F₇O |
CAS-Nummer:
56860-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...