1,1,2,3,3,3-Hexafluorpropylmethylether

Lieferant: FLUOROCHEM

010638-5G 010638-25G
FLUO010638-5GEA 0 EUR
FLUO010638-5G FLUO010638-25G
1,1,2,3,3,3-Hexafluorpropylmethylether
1,1,2,3,3,3-Hexafluorpropylmethylether
Formel: C₄H₄F₆O
MDL: MFCD00041537
CAS-Nummer: 382-34-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR