4-Brom-N,N-diethylanilin ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
L03619.04
L03619.09
L03619.04EA
26.9
EUR
L03619.04
L03619.09
4-Brom-N,N-diethylanilin ≥97%
4-Brom-N,N-diethylanilin
Purity: 97%
CAS No.: 2052-06-4
Molecular Formula: C10H14BrN
Formula Weight: 228.13
Melting Point: 32-35°
Boiling Point: 269-271°
UN No.: 2811
MDL No.: MFCD00013530
Formel:
C₁₀H₁₄BrN Molecular Weight: 228,13 g/mol Siedepunkt: 269…271 °C Schmelzpunkt: 32…35 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013530 CAS-Nummer: 2052-06-4 EINECS: 218-140-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...