1,3-Benzodioxol-4-carbaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Benzo[d][1,3]dioxole-4-carbaldehyde, 2,3-(Methylenedioxy)benzaldehyde, 1,3-Benzodioxole-4-carboxaldehyde

M-Clarity™ Qualitätsstufe = MQ100

389889-1G 389889-5G
SIAL389889-1GEA 44.8 EUR
SIAL389889-1G SIAL389889-5G
1,3-Benzodioxol-4-carbaldehyd, Sigma-Aldrich®
1,3-Benzodioxol-4-carbaldehyd
Formel: C₈H₆O₃
Molecular Weight: 150,1300 g/mol
MDL: MFCD00016902
CAS-Nummer: 7797-83-3
EINECS: 232-252-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR