2,5-Dichlor-4-iodpyridin

Lieferant: COMBI-BLOCKS

SS-3017-1G SS-3017-5G
COBBSS-3017-1GEA 88.9 EUR
COBBSS-3017-1G COBBSS-3017-5G
2,5-Dichlor-4-iodpyridin
2,5-Dichloro-4-iodopyridine
Formel: C₅H₂Cl₂IN
Molecular Weight: 273,8900 g/mol
MDL: MFCD13185538
CAS-Nummer: 796851-03-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR