2,3,5,6-Tetrachlor-4-iodpyridin

Lieferant: Thermo Scientific

BTB15043.250 BTB15043.1
MAYBBTB15043.250EA 0 EUR
MAYBBTB15043.250 MAYBBTB15043.1
2,3,5,6-Tetrachlor-4-iodpyridin
2,3,5,6-Tetrachlor-4-iodpyridin
Formel: C₅Cl₄IN
CAS-Nummer: 30332-35-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR