N-Butylphthalimid ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-Butylisoindoline-1,3-dione, N-(n-Butyl)phthalimide

A16020.06 A16020.14
A16020.06EA 28.5 EUR
A16020.06 A16020.14
N-Butylphthalimid ≥99%
N-Butylphthalimid

Purity: 99% CAS No.: 1515-72-6 Molecular Formula: C12H13NO2 Formula Weight: 203.24 Melting Point: 32-35° Boiling Point: 120-123°/5mm MDL No.: MFCD00039695

Formel: C₁₂H₁₃NO₂
Siedepunkt: 120…123 °C (5 mmHg)
Schmelzpunkt: 32…35 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039695
CAS-Nummer: 1515-72-6
EINECS: 216-157-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR