4-Isopropylphenylessigsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(4-Isopropylphenyl)acetic acid

L05071.03 L05071.06
L05071.03EA 65.9 EUR
L05071.03 L05071.06
4-Isopropylphenylessigsäure ≥98%
4-Isopropylphenylessigsäure

Purity: 98+% CAS No.: 4476-28-2 Molecular Formula: C11H14O2 Formula Weight: 178.23 Melting Point: 51-52° Beilstein Reference: 2328756

Formel: C₁₁H₁₄O₂
Molecular Weight: 178,23 g/mol
Schmelzpunkt: 51…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041035
CAS-Nummer: 4476-28-2
EINECS: 224-755-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR