(4-(Hydroxymethyl)phenyl)essigsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2-(4-(Hydroxymethyl)phenyl)acetic acid, 4-(Hydroxymethyl)phenylacetic acid

M-Clarity™ Qualitätsstufe = MQ100

579610-1G 579610-5G
SIAL579610-1GEA 62.6 EUR
SIAL579610-1G SIAL579610-5G
(4-(Hydroxymethyl)phenyl)essigsäure, Sigma-Aldrich®
(4-(Hydroxymethyl)phenyl)essigsäure
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
MDL: MFCD00065692
CAS-Nummer: 73401-74-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR