4-Fluor-1,3-benzodioxol

Lieferant: COMBI-BLOCKS

Synonyme: 3-Fluro-1,2-methylenedioxybenzene

OT-0855-1G OT-0855-250M
COBBOT-0855-1GEA 623 EUR
COBBOT-0855-1G COBBOT-0855-250M
4-Fluor-1,3-benzodioxol
4-Fluor-1,3-benzodioxol
Formel: C₇H₅FO₂
MDL: MFCD12026062
CAS-Nummer: 943830-74-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR