(5-Fluor-2-methylphenyl)essigsäure

Lieferant: FLUOROCHEM

005806-1G 005806-2.5G
FLUO005806-1GEA 144 EUR
FLUO005806-1G FLUO005806-2.5G
(5-Fluor-2-methylphenyl)essigsäure
(5-Fluor-2-methylphenyl)essigsäure
Formel: C₉H₉FO₂
Molecular Weight: 168,17 g/mol
MDL: MFCD01631530
CAS-Nummer: 261951-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR