1-(Methoxymethyl)-2-(trifluormethyl)benzol

Lieferant: COMBI-BLOCKS

OT-0768-5G OT-0768-1G
COBBOT-0768-5GEA 451 EUR
COBBOT-0768-5G COBBOT-0768-1G
1-(Methoxymethyl)-2-(trifluormethyl)benzol
1-(Methoxymethyl)-2-(trifluoromethyl)benzene
Formel: C₉H₉F₃O
MDL: MFCD11504957
CAS-Nummer: 227948-39-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR