1-(Methoxymethyl)-4-(trifluormethyl)benzol

Lieferant: COMBI-BLOCKS

OT-0770-5G OT-0770-1G
COBBOT-0770-5GEA 595 EUR
COBBOT-0770-5G COBBOT-0770-1G
1-(Methoxymethyl)-4-(trifluormethyl)benzol
1-(Methoxymethyl)-4-(trifluoromethyl)benzene
Formel: C₉H₉F₃O
Molecular Weight: 190,162 g/mol
MDL: MFCD11504959
CAS-Nummer: 155820-05-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR