(±)-β-Methylphenethylamin
Lieferant: FLUOROCHEM
Synonyme:
2-Phenylpropylamine
008888-1G
008888-5G
FLUO008888-1GEA
55.4
EUR
FLUO008888-1G
FLUO008888-5G
(±)-β-Methylphenethylamin
(±)-β-Methylphenethylamin
Formel:
C₉H₁₃N |
MDL:
MFCD00008144 CAS-Nummer: 582-22-9 EINECS: 209-479-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...