1,3,5-Tris(trifluormethyl)benzol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

424765-1G 424765-5G
SIAL424765-1GEA 88.2 EUR
SIAL424765-1G SIAL424765-5G
1,3,5-Tris(trifluormethyl)benzol, Sigma-Aldrich®
1,3,5-Tris(trifluormethyl)benzol
Formel: C₉H₃F₉
Molecular Weight: 282,105 g/mol
MDL: MFCD00054722
CAS-Nummer: 729-81-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR