3-Coumaranon ≥97%
Lieferant: Thermo Scientific
Synonyme:
Benzofuran-3(2H)-one
A10202.03
A10202.06
A10202.14
A10202.03EA
47.3
EUR
A10202.03
A10202.06
A10202.14
3-Coumaranon ≥97%
3-Coumaranon
Purity: 97%
CAS No.: 7169-34-8
Molecular Formula: C8H6O2
Formula Weight: 134.13
Storage Temperature: -30°C to -10°C
Melting Point: 97-102°
Beilstein Reference: 115296
MDL No.: MFCD00051810
Formel:
C₈H₆O₂ Molecular Weight: 134,13 g/mol Schmelzpunkt: 97…102 °C |
MDL:
MFCD00051810 CAS-Nummer: 7169-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...