2-Coumaranon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Benzofuran-2(3H)-one, Benzo[b]furan-2(3H)-one

M-Clarity™ Qualitätsstufe = MQ100

124591-10G 124591-1G
SIAL124591-10GEA 170 EUR
SIAL124591-10G SIAL124591-1G
2-Coumaranon, Sigma-Aldrich®
2-Coumaranon
Formel: C₈H₆O₂
Molecular Weight: 134,13 g/mol
MDL: MFCD00005856
CAS-Nummer: 553-86-6
EINECS: 209-052-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR