Vinylferrocen, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
283630-1G
283630-5G
SIAL283630-1GEA
121
EUR
SIAL283630-1G
SIAL283630-5G
Vinylferrocen, Sigma-Aldrich®
Vinylferrocen
Formel:
C₁₂H₁₂Fe Molecular Weight: 212.07 g/mol |
MDL:
MFCD00001434 CAS-Nummer: 1271-51-8 EINECS: 215-041-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...