Ethyl-3-hydroxy-2-naphthoat
Lieferant: Thermo Scientific
Synonyme:
3-Hydroxy-2-naphthoic acid ethyl ester, Ethyl 3-hydroxy-2-naphthoate
RJC01299.250
RJC01299.1
MAYBRJC01299.250EA
0
EUR
MAYBRJC01299.250
MAYBRJC01299.1
Ethyl-3-hydroxy-2-naphthoat
Ethyl-3-hydroxy-2-naphthoat
Formel:
C₁₃H₁₂O₃ Molecular Weight: 216,2326 g/mol |
MDL:
MFCD00220608 CAS-Nummer: 7163-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...