4,4'-Dibromoctafluorbiphenyl (DBOB)

Lieferant: FLUOROCHEM

Synonyme: 4,4-Dibromooctafluorobiphenyl

009423-5G 009423-25G 009423-1G
FLUO009423-5GEA 266 EUR
FLUO009423-5G FLUO009423-25G FLUO009423-1G
4,4'-Dibromoctafluorbiphenyl (DBOB)
4,4'-Dibromoctafluorbiphenyl (DBOB)
Formel: C₁₂Br₂F₈
Molecular Weight: 455.94 g/mol
MDL: MFCD00000310
CAS-Nummer: 10386-84-2
EINECS: 233-847-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR