trans-11-Methyl-2-dodecensäure

Lieferant: Cayman Chemical

10008321-1 10008321-10 10008321-5
CAYM10008321-1EA 40 EUR
CAYM10008321-1 CAYM10008321-10 CAYM10008321-5
trans-11-Methyl-2-dodecensäure
trans-11-Methyl-2-dodecenoic acid
Formel: C₁₃H₂₄O₂
CAS-Nummer: 677354-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR