3,5-Dimethoxyphenethylamin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

560545-1G 560545-5G
SIAL560545-1GEA 95.9 EUR
SIAL560545-1G SIAL560545-5G
3,5-Dimethoxyphenethylamin, Sigma-Aldrich®
3,5-Dimethoxyphenethylamin
Formel: C₁₀H₁₅NO₂
CAS-Nummer: 3213-28-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR