D(+)-α-Acetobromglucose ≥97% stabilisiert
Lieferant: Thermo Scientific
Synonyme:
2,3,4,6-Tetra-O-acetyl-α-D-glucopyranosyl bromide, Bromo-2,3,4,6-Tetra-o-Acetyl-α-D-Glucopyranose, Acetobromo glucose, (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-bromotetrahydro-2H-pyran-3,4,5-triyl triacetate, α-Acetobromoglucose, Acetobromo-α-D-glucose
Stabilisierung:
Stabilisiert mit Calciumcarbonat
Stabilized with 2% calcium carbonate
Purity: 97%
CAS No.: 572-09-8
Molecular Formula: C14H19BrO9
Formula Weight: 411.22
Storage Temperature: -30°C to -10°C
Melting Point: 84-89°
Beilstein Reference: 96669
Merck Reference: 14,60
Optical Rotation: +198° (c=3 in chloroform)
MDL No.: MFCD00063254
Formel:
C₁₄H₁₉BrO₉ Molecular Weight: 411,2 g/mol Schmelzpunkt: 84…89 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00063254 CAS-Nummer: 572-09-8 EINECS: 209-339-0 Merck Index: 14,60 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...