D(+)-α-Acetobromglucose, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,3,4,6-Tetra-O-acetyl-α-D-glucopyranosyl bromide, Bromo-2,3,4,6-Tetra-o-Acetyl-α-D-Glucopyranose, Acetobromo glucose, (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-bromotetrahydro-2H-pyran-3,4,5-triyl triacetate, α-Acetobromoglucose, Acetobromo-α-D-glucose

M-Clarity™ Qualitätsstufe = MQ300

A1750-100G A1750-10G A1750-25G A1750-50G
SIALA1750-100GEA 687 EUR
SIALA1750-100G SIALA1750-10G SIALA1750-25G SIALA1750-50G
D(+)-α-Acetobromglucose, Sigma-Aldrich®
D(+)-α-Acetobromglucose
Formel: C₁₄H₁₉BrO₉
Molecular Weight: 411,2 g/mol
MDL: MFCD00063254
CAS-Nummer: 572-09-8
EINECS: 209-339-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR