(±)-2-Amino-2-(4-bromphenyl)essigsäure

Lieferant: BIONET RESEARCH

6L-349S.500 6L-349S.1 6L-349S.5 6L-349S.10
BIOT6L-349S.500EA 0 EUR
BIOT6L-349S.500 BIOT6L-349S.1 BIOT6L-349S.5 BIOT6L-349S.10
(±)-2-Amino-2-(4-bromphenyl)essigsäure
(±)-2-Amino-2-(4-bromphenyl)essigsäure
Formel: C₈H₈BrNO₂
CAS-Nummer: 129592-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR