(±)-2-Amino-2-(4-chlorphenyl)essigsäure

Lieferant: BIONET RESEARCH

Synonyme: DL-2-(4-Chlorophenyl)glycine, H-DL-Phg(4-Cl)-OH, DL-alpha-Amino-4-chlorophenylacetic acid, 4-Chloro-DL-2-phenylglycine, 4-Chloro-DL-phenylglycine, 2-Amino-2-(4-chlorophenyl)acetic acid

7L-304S.500 7L-304S.1 7L-304S.5 7L-304S.10
BIOT7L-304S.500EA 0 EUR
BIOT7L-304S.500 BIOT7L-304S.1 BIOT7L-304S.5 BIOT7L-304S.10
(±)-2-Amino-2-(4-chlorphenyl)essigsäure
(±)-2-Amino-2-(4-chlorphenyl)essigsäure
Formel: C₈H₈ClNO₂
Molecular Weight: 185.61 g/mol
MDL: MFCD00049328
CAS-Nummer: 6212-33-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR