1-Naphthoylchlorid

Lieferant: CHEMPUR

FL13172.POR 013172-100G
CHMPFL13172.POREA 0 EUR
CHMPFL13172.POR FLUO013172-100G
1-Naphthoylchlorid
1-Naphthoylchlorid
Formel: C₁₁H₇ClO
Molecular Weight: 190,63 g/mol
MDL: MFCD00004002
CAS-Nummer: 879-18-5
EINECS: 212-903-9
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR