1-Naphthoylchlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

250252-10G 250252-50G
SIAL250252-10GEA 59.1 EUR
SIAL250252-10G SIAL250252-50G
1-Naphthoylchlorid, Sigma-Aldrich®
1-Naphthoylchlorid
Formel: C₁₁H₇ClO
Molecular Weight: 190,63 g/mol
MDL: MFCD00004002
CAS-Nummer: 879-18-5
EINECS: 212-903-9
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR