6-Amino-2-naphthoesäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Amino-2-naphthoic acid, 6-Amino-2-naphthalenecarboxylic acid, 6-Amino-2-naphthoic Acid

M-Clarity™ Qualitätsstufe = MQ100

512370-1G 512370-5G
SIAL512370-1GEA 77 EUR
SIAL512370-1G SIAL512370-5G
6-Amino-2-naphthoesäure, Sigma-Aldrich®
6-Amino-2-naphthoesäure
Formel: C₁₁H₉NO₂
MDL: MFCD01861831
CAS-Nummer: 116668-47-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR