Phloretin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

P7912-100MG P7912-250MG P7912-25MG
SIALP7912-100MGEA 172 EUR
SIALP7912-100MG SIALP7912-250MG SIALP7912-25MG
Phloretin, Sigma-Aldrich®
Phloretin
Formel: C₁₅H₁₄O₅
Molecular Weight: 274,28 g/mol
MDL: MFCD00002288
CAS-Nummer: 60-82-2
EINECS: 200-488-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR