Neodymtrinitrat Hydrat ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®

Lieferant: Thermo Scientific
Gefahr

Synonyme: Neodymium(III) nitrate hydrate

11245.06 11245.14
11245.06EA 44.1 EUR
11245.06 11245.14
Neodymtrinitrat Hydrat ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Neodymtrinitrat Hydrat

Purity: 99.99% CAS No.: 13746-96-8 Molecular Formula: Nd(NO3)3.xH2O Formula Weight: 330.25(anhy) Physical Form: Wet Crystalline Aggregates Sensitivity: Hygroscopic pH: 3.0-4.0 Melting Point: ca 40° Merck Reference: 14,6450 UN No.: 1477 MDL No.: MFCD00149798

Formel: N₃NdO₉·H₂O
Molecular Weight: 330,25 g/mol (anhy)
Schmelzpunkt: ∼ 40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149798
CAS-Nummer: 13746-96-8
EINECS: 233-153-9
UN: 1477
ADR: 5.1,III
Merck Index: 14,6450

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR