2'-Hydroxy-5'-methylpropiophenon

Lieferant: CHEMPUR

FL12086.POR 012086-25G 012086-5G
CHMPFL12086.POREA 0 EUR
CHMPFL12086.POR FLUO012086-25G FLUO012086-5G
2'-Hydroxy-5'-methylpropiophenon
2'-Hydroxy-5'-methylpropiophenon
Formel: C₁₀H₁₂O₂
MDL: MFCD01098830
CAS-Nummer: 938-45-4
EINECS: 213-342-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR